Assay Details
Amplicon Length: | 102 |
Chromosome Location: | Chr. 8 - 140732266 - 140732266 [+] on 140732266 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-151a-3p | MIMAT0000757 |
| Human | hsa-miR-151a-5p | MIMAT0004697 |
Stem-loop Details
Human

Genomic Map