Assay Details
Gene Family ID: | MIPF0000057, mir-28 |
Mature miRNA Sequence: | CUAGACUGAAGCUCCUUGAGG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-151a-3p | MIMAT0000757 |
Bovine | bta-miR-151-3p | MIMAT0003524 |
Dasypus novemcinctus | dno-miR-151-3p | MIMAT0047678 |
Goat | chi-miR-151-3p | MIMAT0035982 |
Pan troglodytes | ptr-miR-151 | MIMAT0008045 |
Pongo pygmaeus | ppy-miR-151a-3p | MIMAT0015776 |
View More Rows | ||
Rabbit | ocu-miR-151-3p | MIMAT0048266 |
Rhesus monkey | mml-miR-151-3p | MIMAT0006213 |
Stem-loop Details
Human
Bovine
Dasypus novemcinctus
Goat
Pan troglodytes
Pongo pygmaeus
Rabbit
Rhesus monkey

More Information
Related Products
miRBase Alias:
Human: hsa-miR-151-3p (v17)
Bos taurus: bta-miR-151 (v18)
Pongo pygmaeus: ppy-miR-151-3p (v18)
Bos taurus: bta-miR-151 (v18)
Pongo pygmaeus: ppy-miR-151-3p (v18)
