Assay Details
Gene Family ID: | MIPF0000287, mir-452 |
Mature miRNA Sequence: | AACUGUUUGCAGAGGAAACUGA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-452-5p | MIMAT0001635 | |
| Callithrix jacchus | cja-miR-452 | MIMAT0039350 | |
| Dog | cfa-miR-452 | MIMAT0009900 | |
| Gorilla gorilla | ggo-miR-452 | MIMAT0024211 | |
| Pan troglodytes | ptr-miR-452 | MIMAT0008154 | |
| Pig | ssc-miR-452 | MIMAT0025374 | |
| View More Rows | |||
| Pongo pygmaeus | ppy-miR-452 | MIMAT0015897 | |
| Pteropus alecto | pal-miR-452-5p | MIMAT0040169 | |
| Rhesus monkey | mml-miR-452-5p | MIMAT0006336 | |
Stem-loop Details
Human
Callithrix jacchus
Dog
Gorilla gorilla
Pan troglodytes
Pig
Pongo pygmaeus
Pteropus alecto
Rhesus monkey
More Information
Related Products
miRBase Alias:
Human: hsa-miR-452 (v17)
Macaca mulatta: mml-miR-452 (v19)
Macaca mulatta: mml-miR-452 (v19)

Genomic Map