Assay Details
Amplicon Length: | 64 |
Chromosome Location: | Chr. X - 151960123 - 151960123 [+] on 151960123 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-452-3p | MIMAT0001636 |
Human | hsa-miR-452-5p | MIMAT0001635 |
Stem-loop Details
Human
