Assay Details
Gene Family ID: | MIPF0000287, mir-452 |
Mature miRNA Sequence: | AACUGUUUGCAGAGGAAACUGA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-452-5p | MIMAT0001635 |
Callithrix jacchus | cja-miR-452 | MIMAT0039350 |
Dog | cfa-miR-452 | MIMAT0009900 |
Gorilla gorilla | ggo-miR-452 | MIMAT0024211 |
Pan troglodytes | ptr-miR-452 | MIMAT0008154 |
Pig | ssc-miR-452 | MIMAT0025374 |
View More Rows | ||
Pongo pygmaeus | ppy-miR-452 | MIMAT0015897 |
Pteropus alecto | pal-miR-452-5p | MIMAT0040169 |
Rhesus monkey | mml-miR-452-5p | MIMAT0006336 |
Stem-loop Details
Human
Callithrix jacchus
Dog
Gorilla gorilla
Pan troglodytes
Pig
Pongo pygmaeus
Pteropus alecto
Rhesus monkey

More Information
Related Products
miRBase Alias:
Human: hsa-miR-452 (v17)
Macaca mulatta: mml-miR-452 (v19)
Macaca mulatta: mml-miR-452 (v19)
