Assay Details
Gene Family ID: | MIPF0000386, mir-743 |
Mature miRNA Sequence: | UAUUCAGAUUAGUGCCAGUCAUG |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-871-5p | MIMAT0004841 |
Stem-loop Details
Mouse

More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Rodent Pool AMegaplex™ RT Primers, Human Pool A
Related Products
miRBase Alias:
Human: hsa-miR-871 (v10)
Mouse: mmu-miR-871 (v15)
Mouse: mmu-miR-871 (v15)
