Assay Details
Gene Family ID: | MIPF0000386, mir-743 |
Mature miRNA Sequence: | UAUUCAGAUUAGUGCCAGUCAUG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-871-5p | MIMAT0004841 |
Stem-loop Details
Mouse

More Information
Related Products
miRBase Alias:
Human: hsa-miR-871 (v10)
Mouse: mmu-miR-871 (v15)
Mouse: mmu-miR-871 (v15)
