Assay Details
Gene Family ID: | MIPF0000020, mir-515 |
Mature miRNA Sequence: | AUCGUGCAUCCCUUUAGAGUGU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-517a-3p | MIMAT0002852 |
| Human | hsa-miR-517b-3p | MIMAT0002857 |
| Gorilla gorilla | ggo-miR-517c-3p | MIMAT0036433 |
| Pan troglodytes | ptr-miR-517a | MIMAT0008184 |
| Pongo pygmaeus | ppy-miR-517a | MIMAT0015948 |
Stem-loop Details
Human
Gorilla gorilla
Pan troglodytes
Pongo pygmaeus
More Information
Related Products
miRBase Alias:
Human: hsa-miR-517a (v17)
Pongo pygmaeus: ppy-miR-517c (v21)
Pongo pygmaeus: ppy-miR-517c (v21)

Genomic Map