Assay Details
| Gene Family ID: | MIPF0000020, mir-515 | 
| Mature miRNA Sequence: | AUCGUGCAUCCCUUUAGAGUGU | 
miRBase Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Human | hsa-miR-517a-3p | MIMAT0002852 | 
| Human | hsa-miR-517b-3p | MIMAT0002857 | 
| Gorilla gorilla | ggo-miR-517c-3p | MIMAT0036433 | 
| Pan troglodytes | ptr-miR-517a | MIMAT0008184 | 
| Pongo pygmaeus | ppy-miR-517a | MIMAT0015948 | 
Stem-loop Details
Human
Gorilla gorilla
Pan troglodytes
Pongo pygmaeus
 Back To Top
 Back To TopMore Information
Related Products
miRBase Alias:
						
							Human: hsa-miR-517a (v17)
Pongo pygmaeus: ppy-miR-517c (v21)
						
					
				Pongo pygmaeus: ppy-miR-517c (v21)
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top TaqMan™ Pri-miRNA Assay :
						TaqMan™ Pri-miRNA Assay :