Assay Details
Gene Family ID: | MIPF0000099, mir-136 |
Mature miRNA Sequence: | ACUCCAUUUGUUUUGAUGAUGG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-136-5p | MIMAT0000148 |
| Eptesicus fuscus | efu-miR-136 | MIMAT0034932 |
| Goat | chi-miR-136-5p | MIMAT0035955 |
| Horse | eca-miR-136 | MIMAT0013128 |
Stem-loop Details
Mouse
Eptesicus fuscus
Goat
Horse

Genomic Map