Assay Details
Gene Family ID: | MIPF0000099, mir-136 |
Mature miRNA Sequence: | ACUCCAUUUGUUUUGAUGAUGG |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-136-5p | MIMAT0000148 |
Eptesicus fuscus | efu-miR-136 | MIMAT0034932 |
Goat | chi-miR-136-5p | MIMAT0035955 |
Horse | eca-miR-136 | MIMAT0013128 |
Stem-loop Details
Mouse
Eptesicus fuscus
Goat
Horse
