Assay Details
Amplicon Length: | 63 |
Chromosome Location: | Chr. 12 - 109595525 - 109595525 [-] on 109595525 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-136-3p | MIMAT0004532 |
| Mouse | mmu-miR-136-5p | MIMAT0000148 |
| Mouse | mmu-miR-432 | MIMAT0012771 |
Stem-loop Details
Mouse

Genomic Map