Assay Details
Gene Family ID: | MIPF0000180, mir-483 |
Mature miRNA Sequence: | UCACUCCUCCCCUCCCGUCUU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-483-3p | MIMAT0003120 |
| Dog | cfa-miR-483 | MIMAT0009901 |
Stem-loop Details
Mouse
Dog

Genomic Map