Assay Details
Amplicon Length: | 71 |
Chromosome Location: | Chr. 7 - 142654726 - 142654726 [+] on 142654726 Build GRCm38 |
Species: | Mouse |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-483-5p | MIMAT0004782 |
Mouse | mmu-miR-483-3p | MIMAT0003120 |
Stem-loop Details
Mouse
