Assay Details
Gene Family ID: | MIPF0000180, mir-483 |
Mature miRNA Sequence: | UCACUCCUCCCCUCCCGUCUU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-483-3p | MIMAT0003120 |
| Dog | cfa-miR-483 | MIMAT0009901 |
Stem-loop Details
Mouse
Dog
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Assay Name: | mmu-miR-483-3p |
|---|---|
| Stem-loop Accession Number: | MI0003484 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UCACUCCUCCCCUCCCGUCUU |
| Species: | Mouse, Dog |
| Product Type: | TaqMan™ Advanced miRNA Assay |
Gene Family ID: | MIPF0000180, mir-483 |
Mature miRNA Sequence: | UCACUCCUCCCCUCCCGUCUU |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-483-3p | MIMAT0003120 |
| Dog | cfa-miR-483 | MIMAT0009901 |