Assay Details
Gene Family ID: | MIPF0000186, mir-299 |
Mature miRNA Sequence: | UAUGUGGGACGGUAAACCGCUU |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-299a-3p | MIMAT0004577 |
Guinea pig | cpo-miR-299-3p | MIMAT0047141 |
Pongo pygmaeus | ppy-miR-299-3p | MIMAT0015814 |
Pteropus alecto | pal-miR-299a-3p | MIMAT0039972 |
Rabbit | ocu-miR-299-3p | MIMAT0048363 |
Rhesus monkey | mml-miR-299-3p | MIMAT0006256 |
Stem-loop Details
Mouse
Guinea pig
Pongo pygmaeus
Pteropus alecto
Rabbit
Rhesus monkey

More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-299-3p (v18)
Mouse: mmu-miR-299 (v17)
Ovis aries: oar-miR-299-3p (v19)
Mouse: mmu-miR-299 (v17)
Ovis aries: oar-miR-299-3p (v19)
