Assay Details
Amplicon Length: | 63 |
Chromosome Location: | Chr. 12 - 109710845 - 109710845 [-] on 109710845 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-299a-5p | MIMAT0000377 |
| Mouse | mmu-miR-299a-3p | MIMAT0004577 |
| Mouse | mmu-miR-299b-5p | MIMAT0022836 |
| Mouse | mmu-miR-299b-3p | MIMAT0022837 |
Stem-loop Details
Mouse

Genomic Map