Assay Details
Gene Family ID: | MIPF0000186, mir-299 |
Mature miRNA Sequence: | UAUGUGGGACGGUAAACCGCUU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-299a-3p | MIMAT0004577 |
| Guinea pig | cpo-miR-299-3p | MIMAT0047141 |
| Pongo pygmaeus | ppy-miR-299-3p | MIMAT0015814 |
| Pteropus alecto | pal-miR-299a-3p | MIMAT0039972 |
| Rabbit | ocu-miR-299-3p | MIMAT0048363 |
| Rhesus monkey | mml-miR-299-3p | MIMAT0006256 |
Stem-loop Details
Mouse
Guinea pig
Pongo pygmaeus
Pteropus alecto
Rabbit
Rhesus monkey

Genomic Map