Assay Details
Gene Family ID: | MIPF0000418, mir-590 |
Mature miRNA Sequence: | UAAUUUUAUGUAUAAGCUAGU |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number | |
---|---|---|---|
Human | hsa-miR-590-3p | MIMAT0004801 | |
Mouse | mmu-miR-590-3p | MIMAT0004896 | |
Dog | cfa-miR-590 | MIMAT0006700 | |
Horse | eca-miR-590-3p | MIMAT0013059 | |
Microcebus murinus | mmr-miR-590 | MIMAT0049174 | |
Pan troglodytes | ptr-miR-590 | MIMAT0008261 | |
View More Rows | |||
Pongo pygmaeus | ppy-miR-590-3p | MIMAT0016033 |
Stem-loop Details
Human
Mouse
Dog
Horse
Microcebus murinus
Pan troglodytes
Pongo pygmaeus

More Information
Associated Megaplex™ RT Primer Pools:
Megaplex™ RT Primers, Rodent Pool B v3.0Megaplex™ RT Primers, Human Pool B v3.0
Associated TaqMan® Array MicroRNA Cards:
TaqMan® Human MicroRNA B Cards v3.0TaqMan® Rodent MicroRNA B Cards v3.0
TaqMan® OpenArray® MicroRNA Panels:
TaqMan® OpenArray® Human MicroRNA PanelTaqMan® OpenArray® Rodent MicroRNA Panel
Related Products
