Assay Details
Gene Family ID: | MIPF0000418, mir-590 |
Mature miRNA Sequence: | UAAUUUUAUGUAUAAGCUAGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-590-3p | MIMAT0004801 |
| Mouse | mmu-miR-590-3p | MIMAT0004896 |
| Dog | cfa-miR-590 | MIMAT0006700 |
| Horse | eca-miR-590-3p | MIMAT0013059 |
| Microcebus murinus | mmr-miR-590 | MIMAT0049174 |
| Pan troglodytes | ptr-miR-590 | MIMAT0008261 |
| View More Rows | ||
| Pongo pygmaeus | ppy-miR-590-3p | MIMAT0016033 |
Stem-loop Details
Human
Mouse
Dog
Horse
Microcebus murinus
Pan troglodytes
Pongo pygmaeus

Genomic Map