Assay Details
Gene Family ID: | MIPF0000596, mir-1207 |
Mature miRNA Sequence: | UCAGCUGGCCCUCAUUUC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1207-3p | MIMAT0005872 |
| Pan troglodytes | ptr-miR-1207 | MIMAT0007961 |
| Pongo pygmaeus | ppy-miR-1207-3p | MIMAT0016156 |
Stem-loop Details
Human
Pan troglodytes
Pongo pygmaeus

Genomic Map