Assay Details
Amplicon Length: | 62 |
Chromosome Location: | Chr. 8 - 128049445 - 128049445 [-] on 128049445 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1207-3p | MIMAT0005872 |
| Human | hsa-miR-1207-5p | MIMAT0005871 |
Stem-loop Details
Human

Genomic Map