Assay Details
| Gene Family ID: | MIPF0000596, mir-1207 | 
| Mature miRNA Sequence: | UCAGCUGGCCCUCAUUUC | 
miRBase Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Human | hsa-miR-1207-3p | MIMAT0005872 | 
| Pan troglodytes | ptr-miR-1207 | MIMAT0007961 | 
| Pongo pygmaeus | ppy-miR-1207-3p | MIMAT0016156 | 
Stem-loop Details
Human
Pan troglodytes
Pongo pygmaeus
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top TaqMan™ Pri-miRNA Assay :
						TaqMan™ Pri-miRNA Assay :