Assay Details
Gene Family ID: | MIPF0000027, mir-23 |
Mature miRNA Sequence: | GGGUUCCUGGCAUGCUGAUUU |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Mouse | mmu-miR-23b-5p | MIMAT0016980 | |
| Rat | rno-miR-23b-5p | MIMAT0017099 | |
| Alligator mississippiensis | ami-miR-23b-5p | MIMAT0038059 | |
| Anolis carolinensis | aca-miR-23b-5p | MIMAT0021897 | |
| Bovine | bta-miR-23b-5p | MIMAT0012538 | |
| Dasypus novemcinctus | dno-miR-23b-5p | MIMAT0047530 | |
| View More Rows | |||
| Guinea pig | cpo-miR-23b-5p | MIMAT0046878 | |
| Monodelphis domestica | mdo-miR-23b-5p | MIMAT0026713 | |
| Pteropus alecto | pal-miR-23b-5p | MIMAT0039870 | |
| Python bivittatus | pbv-miR-23b-5p | MIMAT0038854 | |
| Rabbit | ocu-miR-23b-5p | MIMAT0048128 | |
| Rhesus monkey | mml-miR-23b-5p | MIMAT0026800 | |
Stem-loop Details
Mouse
Rat
Alligator mississippiensis
Anolis carolinensis
Bovine
Dasypus novemcinctus
Guinea pig
Monodelphis domestica
Pteropus alecto
Python bivittatus
Rabbit
Rhesus monkey
More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-23b* (v17)
Rat: rno-miR-23b* (v18)
Aeromonas caviae Ae398: aca-miR-23b* (v18)
Rat: rno-miR-23b* (v18)
Aeromonas caviae Ae398: aca-miR-23b* (v18)

Genomic Map