Assay Details
Gene Family ID: | MIPF0000027, mir-23 |
Mature miRNA Sequence: | GGGUUCCUGGCAUGCUGAUUU |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-23b-5p | MIMAT0016980 |
Rat | rno-miR-23b-5p | MIMAT0017099 |
Alligator mississippiensis | ami-miR-23b-5p | MIMAT0038059 |
Anolis carolinensis | aca-miR-23b-5p | MIMAT0021897 |
Bovine | bta-miR-23b-5p | MIMAT0012538 |
Dasypus novemcinctus | dno-miR-23b-5p | MIMAT0047530 |
View More Rows | ||
Guinea pig | cpo-miR-23b-5p | MIMAT0046878 |
Monodelphis domestica | mdo-miR-23b-5p | MIMAT0026713 |
Pteropus alecto | pal-miR-23b-5p | MIMAT0039870 |
Python bivittatus | pbv-miR-23b-5p | MIMAT0038854 |
Rabbit | ocu-miR-23b-5p | MIMAT0048128 |
Rhesus monkey | mml-miR-23b-5p | MIMAT0026800 |
Stem-loop Details
Mouse
Rat
Alligator mississippiensis
Anolis carolinensis
Bovine
Dasypus novemcinctus
Guinea pig
Monodelphis domestica
Pteropus alecto
Python bivittatus
Rabbit
Rhesus monkey

More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-23b* (v17)
Rat: rno-miR-23b* (v18)
Aeromonas caviae Ae398: aca-miR-23b* (v18)
Rat: rno-miR-23b* (v18)
Aeromonas caviae Ae398: aca-miR-23b* (v18)
