Assay Details
Gene Family ID: | MIPF0000180, mir-483 |
Mature miRNA Sequence: | CACUCCUCCCCUCCCGUCUUGU |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-483-3p | MIMAT0003121 |
Guinea pig | cpo-miR-483-3p | MIMAT0047274 |
Stem-loop Details
Rat
Guinea pig
