Assay Details
| Amplicon Length: | 74 | 
| Chromosome Location: | Chr. 1 - 215831840 - 215831840 [+] on 215831840 Build Rnor_6.0 | 
| Species: | Rat | 
miRBase Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Rat | rno-miR-483-5p | MIMAT0017201 | 
| Rat | rno-miR-483-3p | MIMAT0003121 | 
Stem-loop Details
Rat
 Back To Top
 Back To Top
 rno-miR-483-5p
 rno-miR-483-5p Genomic Map
Genomic Map Back To Top
 Back To Top Ambion® Anti-miR™ miRNA Inhibitor :
						Ambion® Anti-miR™ miRNA Inhibitor :