Assay Details
| Gene Family ID: | MIPF0000180, mir-483 | 
| Mature miRNA Sequence: | CACUCCUCCCCUCCCGUCUUGU | 
miRBase Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Rat | rno-miR-483-3p | MIMAT0003121 | 
| Guinea pig | cpo-miR-483-3p | MIMAT0047274 | 
Stem-loop Details
Rat
Guinea pig
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top TaqMan™ Pri-miRNA Assay :
						TaqMan™ Pri-miRNA Assay :