Assay Details
Gene Family ID: | MIPF0000188, mir-346 |
Mature miRNA Sequence: | AGGCAGGGGCUGGGCCUGCAGC |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-346-3p | MIMAT0017039 |
| Guinea pig | cpo-miR-346-3p | MIMAT0047185 |
| Pteropus alecto | pal-miR-346-3p | MIMAT0040342 |
Stem-loop Details
Mouse
Guinea pig
Pteropus alecto

Genomic Map