Assay Details
Gene Family ID: | MIPF0000188, mir-346 | 
Mature miRNA Sequence: | AGGCAGGGGCUGGGCCUGCAGC | 
miRBase Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Mouse | mmu-miR-346-3p | MIMAT0017039 | 
| Guinea pig | cpo-miR-346-3p | MIMAT0047185 | 
| Pteropus alecto | pal-miR-346-3p | MIMAT0040342 | 
Stem-loop Details
Mouse
Guinea pig
Pteropus alecto

Genomic Map