Assay Details
Amplicon Length: | 57 |
Chromosome Location: | Chr. 14 - 34895013 - 34895013 [-] on 34895013 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-346-3p | MIMAT0017039 |
| Mouse | mmu-miR-346-5p | MIMAT0000597 |
Stem-loop Details
Mouse

Genomic Map