Assay Details
Gene Family ID: | MIPF0000126, mir-379 |
Mature miRNA Sequence: | UGGUUGACCAGAGAGCACACG |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-758-5p | MIMAT0017235 |
| Rat | rno-miR-758-5p | MIMAT0017316 |
| Sheep | oar-miR-758-5p | MIMAT0019261 |
Stem-loop Details
Mouse
Rat
Sheep
More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-758* (v17)
Rat: rno-miR-758* (v18)
Rat: rno-miR-758* (v18)

Genomic Map