Assay Details
Amplicon Length: | 72 |
Chromosome Location: | Chr. 12 - 109713211 - 109713211 [-] on 109713211 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-329-5p | MIMAT0017032 |
| Mouse | mmu-miR-329-3p | MIMAT0000567 |
| Mouse | mmu-miR-758-5p | MIMAT0017235 |
| Mouse | mmu-miR-758-3p | MIMAT0003889 |
Stem-loop Details
Mouse

Genomic Map