Assay Details
Amplicon Length: | 74 |
Chromosome Location: | Chr. 6 - 133861049 - 133861049 [-] on 133861049 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-323-3p | MIMAT0000550 |
| Rat | rno-miR-323-5p | MIMAT0004637 |
| Rat | rno-miR-758-3p | MIMAT0005335 |
| Rat | rno-miR-758-5p | MIMAT0017316 |
Stem-loop Details
Rat

Genomic Map