Assay Details
Gene Family ID: | MIPF0000174, mir-454 |
Mature miRNA Sequence: | UAGUGCAAUAUUGCUUAUAGGGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-454-3p | MIMAT0003885 |
| Anolis carolinensis | aca-miR-454-3p | MIMAT0021963 |
| Bovine | bta-miR-454 | MIMAT0009326 |
| Chicken | gga-miR-454-3p | MIMAT0007292 |
| Gadus morhua | gmo-miR-454-3p | MIMAT0044282 |
| Goat | chi-miR-454-3p | MIMAT0036240 |
| View More Rows | ||
| Horse | eca-miR-454 | MIMAT0013042 |
| Ictalurus punctatus | ipu-miR-454b | MIMAT0029511 |
| Nomascus leucogenys | nle-miR-454 | MIMAT0049398 |
| Ornithorhynchus anatinus | oan-miR-454-3p | MIMAT0007157 |
| Pan troglodytes | ptr-miR-454 | MIMAT0008156 |
| Pig | ssc-miR-454 | MIMAT0041609 |
| Rhesus monkey | mml-miR-454-3p | MIMAT0006338 |
| Sarcophilus harrisii | sha-miR-454 | MIMAT0022815 |
| Taeniopygia guttata | tgu-miR-454-3p | MIMAT0014580 |
| Xenopus tropicalis | xtr-miR-454-3p | MIMAT0037590 |
Stem-loop Details
Human
Anolis carolinensis
Bovine
Chicken
Gadus morhua
Goat
Horse
Ictalurus punctatus
Nomascus leucogenys
Ornithorhynchus anatinus
Pan troglodytes
Pig
Rhesus monkey
Sarcophilus harrisii
Taeniopygia guttata
Xenopus tropicalis

Genomic Map