Assay Details
Gene Family ID: | MIPF0000174, mir-454 |
Mature miRNA Sequence: | UAGUGCAAUAUUGCUUAUAGGGU |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-454-3p | MIMAT0003885 |
| Anolis carolinensis | aca-miR-454-3p | MIMAT0021963 |
| Bovine | bta-miR-454 | MIMAT0009326 |
| Chicken | gga-miR-454-3p | MIMAT0007292 |
| Gadus morhua | gmo-miR-454-3p | MIMAT0044282 |
| Goat | chi-miR-454-3p | MIMAT0036240 |
| View More Rows | ||
| Horse | eca-miR-454 | MIMAT0013042 |
| Ictalurus punctatus | ipu-miR-454b | MIMAT0029511 |
| Nomascus leucogenys | nle-miR-454 | MIMAT0049398 |
| Ornithorhynchus anatinus | oan-miR-454-3p | MIMAT0007157 |
| Pan troglodytes | ptr-miR-454 | MIMAT0008156 |
| Pig | ssc-miR-454 | MIMAT0041609 |
| Rhesus monkey | mml-miR-454-3p | MIMAT0006338 |
| Sarcophilus harrisii | sha-miR-454 | MIMAT0022815 |
| Taeniopygia guttata | tgu-miR-454-3p | MIMAT0014580 |
| Xenopus tropicalis | xtr-miR-454-3p | MIMAT0037590 |
Stem-loop Details
Human
Anolis carolinensis
Bovine
Chicken
Gadus morhua
Goat
Horse
Ictalurus punctatus
Nomascus leucogenys
Ornithorhynchus anatinus
Pan troglodytes
Pig
Rhesus monkey
Sarcophilus harrisii
Taeniopygia guttata
Xenopus tropicalis
More Information
Related Products
miRBase Alias:
Human: hsa-miR-454 (v17)
Aeromonas caviae Ae398: aca-miR-454 (v18)
Gallus gallus: gga-miR-454 (v18)
Macaca mulatta: mml-miR-454 (v19)
Ochrobactrum anthropi: oan-miR-454 (v18)
Trabulsiella guamensis ATCC 49490: tgu-miR-454 (v19)
Aeromonas caviae Ae398: aca-miR-454 (v18)
Gallus gallus: gga-miR-454 (v18)
Macaca mulatta: mml-miR-454 (v19)
Ochrobactrum anthropi: oan-miR-454 (v18)
Trabulsiella guamensis ATCC 49490: tgu-miR-454 (v19)

Genomic Map