Assay Details
Amplicon Length: | 100 |
Chromosome Location: | Chr. 17 - 59138048 - 59138048 [-] on 59138048 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-454-5p | MIMAT0003884 |
| Human | hsa-miR-454-3p | MIMAT0003885 |
Stem-loop Details
Human

Genomic Map