Assay Details
Gene Family ID: | MIPF0000709, mir-761 |
Mature miRNA Sequence: | GCAGCAGGGUGAAACUGACACA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-761 | MIMAT0010364 |
| Mouse | mmu-miR-761 | MIMAT0003893 |
| Rat | rno-miR-761 | MIMAT0012853 |
| Bovine | bta-miR-761 | MIMAT0009373 |
| Callithrix jacchus | cja-miR-761 | MIMAT0039612 |
| Dog | cfa-miR-761 | MIMAT0009936 |
| View More Rows | ||
| Horse | eca-miR-761 | MIMAT0012923 |
| Pan troglodytes | ptr-miR-761 | MIMAT0012817 |
| Rhesus monkey | mml-miR-761 | MIMAT0012793 |
Stem-loop Details
Human
Mouse
Rat
Bovine
Callithrix jacchus
Dog
Horse
Pan troglodytes
Rhesus monkey

Genomic Map