Assay Details
Gene Family ID: | MIPF0000709, mir-761 |
Mature miRNA Sequence: | GCAGCAGGGUGAAACUGACACA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-761 | MIMAT0010364 |
Mouse | mmu-miR-761 | MIMAT0003893 |
Rat | rno-miR-761 | MIMAT0012853 |
Bovine | bta-miR-761 | MIMAT0009373 |
Callithrix jacchus | cja-miR-761 | MIMAT0039612 |
Dog | cfa-miR-761 | MIMAT0009936 |
View More Rows | ||
Horse | eca-miR-761 | MIMAT0012923 |
Pan troglodytes | ptr-miR-761 | MIMAT0012817 |
Rhesus monkey | mml-miR-761 | MIMAT0012793 |
Stem-loop Details
Human
Mouse
Rat
Bovine
Callithrix jacchus
Dog
Horse
Pan troglodytes
Rhesus monkey
