Assay Details
Amplicon Length: | 119 |
Chromosome Location: | Chr. 5 - 128651718 - 128651718 [+] on 128651718 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-761 | MIMAT0012853 |
Stem-loop Details
Rat
