Assay Details
Gene Family ID: | MIPF0001064, mir-1304 |
Mature miRNA Sequence: | UCUCACUGUAGCCUCGAACCCC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1304-3p | MIMAT0022720 |
| Gorilla gorilla | ggo-miR-1304 | MIMAT0049120 |
| Pan paniscus | ppa-miR-1304 | MIMAT0049088 |
Stem-loop Details
Human
Gorilla gorilla
Pan paniscus

Genomic Map