Assay Details
Amplicon Length: | 111 |
Chromosome Location: | Chr. 11 - 93734167 - 93734167 [+] on 93734167 Build GRCh38 |
Species: | Human |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1304-5p | MIMAT0005892 |
| Human | hsa-miR-1304-3p | MIMAT0022720 |
Stem-loop Details
Human

Genomic Map