Assay Details
Gene Family ID: | MIPF0001064, mir-1304 | 
Mature miRNA Sequence: | UCUCACUGUAGCCUCGAACCCC | 
miRBase Details:
| Species | miRBase ID | miRBase Accession Number | 
|---|---|---|
| Human | hsa-miR-1304-3p | MIMAT0022720 | 
| Gorilla gorilla | ggo-miR-1304 | MIMAT0049120 | 
| Pan paniscus | ppa-miR-1304 | MIMAT0049088 | 
Stem-loop Details
Human
Gorilla gorilla
Pan paniscus

Genomic Map