Assay Details
Gene Family ID: | MIPF0000018, mir-154 |
Mature miRNA Sequence: | UAUGCAAGGGCAAGCUCUCUUC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-300-3p | MIMAT0000378 |
| Rat | rno-miR-300-3p | MIMAT0000902 |
| Cricetulus griseus | cgr-miR-300 | MIMAT0023889 |
Stem-loop Details
Mouse
Rat
Cricetulus griseus
More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-300 (v17)
Rat: rno-miR-300 (v9.2)
Rat: rno-miR-300 (v9.2)

Genomic Map