Assay Details
Amplicon Length: | 80 |
Chromosome Location: | Chr. 6 - 133873995 - 133873995 [+] on 133873995 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-300-3p | MIMAT0000902 |
Rat | rno-miR-300-5p | MIMAT0004743 |
Stem-loop Details
Rat
