Assay Details
Amplicon Length: | 98 |
Chromosome Location: | Chr. 12 - 109724107 - 109724107 [-] on 109724107 Build GRCm38 |
Species: | Mouse |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Mouse | mmu-miR-300-5p | MIMAT0004578 |
Mouse | mmu-miR-300-3p | MIMAT0000378 |
Stem-loop Details
Mouse
