Assay Details
Gene Family ID: | MIPF0000318, mir-488 |
Mature miRNA Sequence: | UUGAAAGGCUGUUUCUUGGUC |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-488-3p | MIMAT0003450 |
| Rat | rno-miR-488-3p | MIMAT0005341 |
| Bovine | bta-miR-488 | MIMAT0009330 |
Stem-loop Details
Mouse
Rat
Bovine
More Information
Related Products
miRBase Alias:
Mouse: mmu-miR-488 (v17)
Mouse: mmu-miR-488* (v9.2)
Rat: rno-miR-488 (v18)
Mouse: mmu-miR-488* (v9.2)
Rat: rno-miR-488 (v18)

Genomic Map