Assay Details
Amplicon Length: | 52 |
Chromosome Location: | Chr. 13 - 76198442 - 76198442 [-] on 76198442 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-488-3p | MIMAT0005341 |
| Rat | rno-miR-488-5p | MIMAT0017320 |
Stem-loop Details
Rat

Genomic Map