Assay Details
Gene Family ID: | MIPF0000628, mir-1208 |
Mature miRNA Sequence: | UCACUGUUCAGACAGGCGGA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1208 | MIMAT0005873 |
| Pan troglodytes | ptr-miR-1208 | MIMAT0007962 |
| Pongo pygmaeus | ppy-miR-1208 | MIMAT0016157 |
Stem-loop Details
Human
Pan troglodytes
Pongo pygmaeus

Genomic Map