Assay Details
Gene Family ID: | MIPF0000628, mir-1208 |
Mature miRNA Sequence: | UCACUGUUCAGACAGGCGGA |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-1208 | MIMAT0005873 |
Pan troglodytes | ptr-miR-1208 | MIMAT0007962 |
Pongo pygmaeus | ppy-miR-1208 | MIMAT0016157 |
Stem-loop Details
Human
Pan troglodytes
Pongo pygmaeus
